MilliporeSigma
  • Genotyping of human alcohol dehydrogenases at the ADH2 and ADH3 loci following DNA sequence amplification.
Genotyping of human alcohol dehydrogenases at the ADH2 and ADH3 loci following DNA sequence amplification.
Genomics (1988-04-01)
Y L Xu, L G Carr, W F Bosron, T K Li, H J Edenberg
ABSTRACT
Humans are polymorphic at two of the alcohol dehydrogenase (ADH) loci important in ethanol metabolism, ADH2 and ADH3. Although the coding regions of these genes are 94% identical, they produce subunits that differ greatly in kinetic properties in vitro. These differences are likely to be reflected in the pharmacokinetics of alcohol metabolism, but studies have been hampered by the need to use liver biopsy specimens to determine the ADH phenotype. This problem has now been overcome by determining the genotype at these loci using DNA that has been amplified in vitro by the polymerase chain reaction. We report here the identification of all three of the ADH2 alleles and both of the ADH3 alleles. Any pair of ADH2 or ADH3 alleles can be distinguished using allele-specific oligonucleotide probes directed at their single base pair difference. In addition, ADH2(2) can be distinguished from ADH2(1) and ADH2(3) by detecting a new MaeIII site created in the third exon by the single base pair alteration in ADH2(2).
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg