MilliporeSigma
  • Design and synthesis of reagents for phage display screening of dehalogenases.
Design and synthesis of reagents for phage display screening of dehalogenases.
Bioorganic & medicinal chemistry letters (1999-02-18)
R J Pieters, M Fennema, R M Kellogg, D B Janssen
ABSTRACT
Bifunctional molecules containing both a biotin and a substrate unit have been designed and synthesized for phage display screening of mutant libraries of haloalkane dehalogenase enzymes. The molecules were assembled using a convergent modular synthetic strategy. One molecule was synthesized to evaluate the concept of covalent capture and a second for screening of phage libraries for enantioselectivity.
Sigma-Aldrich
1-Bromohexane, 98%