MilliporeSigma
  • Solution structure of the epithelial cadherin domain responsible for selective cell adhesion.
Solution structure of the epithelial cadherin domain responsible for selective cell adhesion.
Science (New York, N.Y.) (1995-01-20)
M Overduin, T S Harvey, S Bagby, K I Tong, P Yau, M Takeichi, M Ikura
ABSTRACT
Cadherins are calcium-dependent cell adhesion molecules containing extracellular repeats of approximately 110 amino acids. The three-dimensional structure of the amino-terminal repeat of mouse epithelial cadherin was determined by multidimensional heteronuclear magnetic resonance spectroscopy. The calcium ion was bound by a short alpha helix and by loops at one end of the seven-stranded beta-barrel structure. An exposed concave face is in a position to provide homophilic binding specificity and was also sensitive to calcium ligation. Unexpected structural similarities with the immunoglobulin fold suggest an evolutionary relation between calcium-dependent and calcium-independent cell adhesion molecules.
Sigma-Aldrich
E-Cadherin/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder