MilliporeSigma
  • Method for the analysis of triadimefon and ethofumesate from dislodgeable foliar residues on turfgrass by solid-phase extraction and in-vial elution.
Method for the analysis of triadimefon and ethofumesate from dislodgeable foliar residues on turfgrass by solid-phase extraction and in-vial elution.
Journal of agricultural and food chemistry (1999-12-20)
H B Runes, J J Jenkins, J A Field
ABSTRACT
Triadimefon, a fungicide, and ethofumesate, an herbicide, are commonly applied to turfgrass in the Pacific Northwest, resulting in foliar residues. A simple and rapid method was developed to determine triadimefon and ethofumesate concentrations from dislodgeable foliar residues on turfgrass. Turfgrass samples were washed, and wash water containing surfactant (a 0.126% solution) was collected for residue analysis. This analytical method utilizes a 25 mm C(8) Empore disk and in-vial elution to quantitatively determine triadimefon and ethofumesate in 170 mL aqueous samples. The analytes were eluted by placing the disk in a 2 mL autosampler vial with 980 microL of ethyl acetate and 20 microL of 2-chlorolepidine, the internal standard, for analysis by GC/MS. The method quantitation limits are 0.29 microg/L for ethofumesate and 0.59 microg/L for triadimefon. The method detection limits are 0.047 microg/L and 0.29 microg/L for ethofumesate and triadimefon, respectively. Concentrations of triadimefon and ethofumesate from dislodgeable foliar residues from a field study are reported.
Product Description
Supelco
Ethofumesate, PESTANAL®, analytical standard