MilliporeSigma
  • Production of polyclonal antibodies towards the immunodetection of insecticide phosalone.
Production of polyclonal antibodies towards the immunodetection of insecticide phosalone.
Amino acids (2000-03-09)
V Issert, R Lazaro, F Lamaty, V Rolland, P Besançon, B Caporiccio, P Grenier, V Bellon-Maurel
ABSTRACT
Hapten synthesis for the production of specific insecticide phosalone polyclonal antibodies was carried out starting from an intermediate of the phosalone synthesis, 6-chloro-2-benzoxazolone 1. Two haptens containing different spacers have been prepared: N-5-carboxypentyl-6-chloro-2-benzoxazolone 7 and N-(2-oxo-3-aza-5-carboxypentyl)-6-chloro-2-benzoxazolone 12. Each of these two haptens conjugated to bovine serum albumine (BSA) was used to immunize four rabbits. Immunoassays of phosalone were performed with ELISA using solid-phase bound hapten thyroglobulin conjugate and horseradish peroxidase labelled goat antirabbit IgG. The more sensitive response was observed when the antiserum obtained from the rabbit immunized with the hapten-BSA conjugate containing the N-2-oxo-3-aza-5-carboxypentyl spacer was in competition with the same hapten coupled to thyroglobulin. An identical response was obtained under the same conditions when using benzoxazolone instead of phosalone as competitor, showing a good recognition of the specific aromatic part of the organophosphate insecticide phosalone. Reduction of coating conjugate concentration (from 2 to 0.05 micrograms/well) and also the use of heterologous coating protein instead of homologous did improve the sensitivity, resulting in a concentration of phosalone required to inhibit binding by 50% of 2 mg/l and a detection limit of 0.02 mg/l.
Product Description
Supelco
Phosalone, PESTANAL®, analytical standard