MilliporeSigma
  • Design of dimeric peptides obtained from a subdominant Epstein-Barr virus LMP2-derived epitope.
Design of dimeric peptides obtained from a subdominant Epstein-Barr virus LMP2-derived epitope.
European journal of medicinal chemistry (2000-07-25)
M Marastoni, M Bazzaro, R Gavioli, F Micheletti, S Traniello, R Tomatis
ABSTRACT
The latent membrane protein 2 (LMP2) is expressed in EBV-associated tumours. LMP2 is a target of HLA-A2 restricted EBV-specific CTL responses and consequently it may represent a good target for specific CTL-based immunotherapies. However, the efficacy of such therapy is limited by the poor immunogenicity of the protein that induces weak cytotoxic T lymphocyte (CTL) responses directed against the CLGGLLTMV (CLG) epitope. Indeed, the CLG peptide presents low affinity for HLA-A2 and does not produce stable complexes. Therefore we synthesized and tested CLG-dimeric analogues with the purpose of characterizing new compounds with the capacity to bind HLA-A2 molecules. By these studies we have identified a few peptides which, compared to the natural epitope, showed higher affinity for HLA-A2 molecules and superior capacity to form a complex. These dimeric peptides may have the potential to induce efficient CTL responses directed to the natural epitope.
Sigma-Aldrich
11-Aminoundecanoic acid, 97%