MilliporeSigma
  • Single-crystalline ceramic whisker-reinforced carboxylic acid-resin composites with fluoride release.
Single-crystalline ceramic whisker-reinforced carboxylic acid-resin composites with fluoride release.
Operative dentistry (2001-02-24)
H H Xu, F C Eichmiller, J M Antonucci, G M Flaim
ABSTRACT
Currently available glass-ionomer, resin-modified glass-ionomer, and compomer materials have relatively low strength and toughness and, therefore, are inadequate for use in large stress-bearing posterior restorations. In the present study, ceramic single-crystalline whiskers were mixed with fluorosilicate glass particles and used as fillers to reinforce experimental carboxylic acid-resin composites. The carboxylic acid was a monofunctional methacryloxyethyl phthalate (MEP). Five mass fractions of whisker/(whisker + fluorosilicate glass), and corresponding resin (resin + MEP), were evaluated. Four control materials were also tested for comparison: a glass ionomer, a resin-modified glass ionomer, a compomer, and a hybrid composite resin. Flexural specimens were fabricated to measure the flexural strength, elastic modulus, and work-of-fracture (an indication of toughness). Fluoride release was measured by using a fluoride ion selective electrode. The properties of whisker composites depended on the whisker/(whisker + fluorosilicate glass) mass fraction. At a mass fraction of 0.8, the whisker composite had a flexural strength in MPa (mean +/- sd; n = 6) of 150 +/- 16, significantly higher than that of a glass ionomer (15 +/- 7) or a compomer control (89 +/- 18) (Tukey's multiple comparison test; family confidence coefficient = 0.95). Depending on the ratio of whisker:fluorosilicate glass, the whisker composites had a cumulative fluoride release up to 60% of that of a traditional glass ionomer. To conclude, combining ceramic whiskers and fluorosilicate glass in a carboxylic acid-resin matrix can result in fluoride-releasing composites with significantly improved mechanical properties.
Sigma-Aldrich
Fluosilicic acid, purum, 33.5-35%
Sigma-Aldrich
Fluorosilicic acid solution, 20-25 wt. % in H2O