Merck
  • Beta1-adrenergic agonist is a potent stimulator of alveolar fluid clearance in hyperoxic rat lungs.
Beta1-adrenergic agonist is a potent stimulator of alveolar fluid clearance in hyperoxic rat lungs.
Japanese journal of pharmacology (2001-04-05)
T Sakuma, M Hida, Y Nambu, K Osanai, H Toga, K Takahashi, N Ohya, M Inoue, Y Watanabe
ABSTRACT
Because it was still uncertain whether a stimulation of beta1-adrenoceptors accelerated alveolar fluid clearance in hyperoxic lung injury, the effect of denopamine, a selective beta1-adrenergic agonist, on alveolar fluid clearance was determined in rats exposed to 93% oxygen for 48 and 56 h. Alveolar fluid clearance was measured by the progressive increase in the concentration of Evans blue labeled albumin instilled into the alveolar spaces over 1 h at 37 degrees C in isolated rat lungs. The principle results were as follows: 1) Although lung water volume increased in rats exposed to hyperoxia for 48 and 56 h, basal alveolar fluid clearance did not change for up to 56 h; 2) Denopamine increased alveolar fluid clearance in rats exposed to hyperoxia as well as in rats without exposure to hyperoxia; 3) Denopamine primarily increased amiloride-insensitive alveolar fluid clearance in rats exposed to hyperoxia; 4) The potency of denopmaine was similar to that of terbutaline, a selective beta2-adrenergic agonist. In summary, denopamine is a potent stimulator of alveolar fluid clearance in rats exposed to hyperoxia.
Sigma-Aldrich
R(−)-Denopamine, ≥98% (HPLC), powder