MilliporeSigma
  • Differential ligand activation of estrogen receptors ERalpha and ERbeta at AP1 sites.
Differential ligand activation of estrogen receptors ERalpha and ERbeta at AP1 sites.
Science (New York, N.Y.) (1997-09-05)
K Paech, P Webb, G G Kuiper, S Nilsson, J Gustafsson, P J Kushner, T S Scanlan
ABSTRACT
The transactivation properties of the two estrogen receptors, ERalpha and ERbeta, were examined with different ligands in the context of an estrogen response element and an AP1 element. ERalpha and ERbeta were shown to signal in opposite ways when complexed with the natural hormone estradiol from an AP1 site: with ERalpha, 17beta-estradiol activated transcription, whereas with ERbeta, 17beta-estradiol inhibited transcription. Moreover, the antiestrogens tamoxifen, raloxifene, and Imperial Chemical Industries 164384 were potent transcriptional activators with ERbeta at an AP1 site. Thus, the two ERs signal in different ways depending on ligand and response element. This suggests that ERalpha and ERbeta may play different roles in gene regulation.
Sigma-Aldrich
Estrogen Receptor-α human, ≥80% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution