MilliporeSigma
  • Synthetic peptides that inhibit binding of the myelin basic protein 85-99 epitope to multiple sclerosis-associated HLA-DR2 molecules and MBP-specific T-cell responses.
Synthetic peptides that inhibit binding of the myelin basic protein 85-99 epitope to multiple sclerosis-associated HLA-DR2 molecules and MBP-specific T-cell responses.
Human immunology (2001-07-31)
M Fridkis-Hareli, J N Stern, L Fugger, J L Strominger
ABSTRACT
Copolymer 1 (Cop 1, poly [Y, E, A, K]) is a random synthetic amino acid copolymer effective in the treatment of relapsing forms of multiple sclerosis (MS), a disease that is linked to HLA-DR2 (DRB1*1501). In the present study various peptides, synthesized according to the binding motifs for both the immunodominant epitope of myelin basic protein (MBP) 85-99, a candidate autoantigen in MS, and Cop 1, differentially inhibited binding of these antigens to disease-associated HLA-DR2 (DRB1*1501) molecules. In particular, two peptides with residue K at position P-1, as referred to MBP 85-99, inhibited effectively the binding of both biotinylated MBP 85-99 and Cop 1 to HLA-DR2 molecules as well as IL-2 production by two MBP-specific HLA-DR2-restricted T-cell clones. These findings suggest the possible utility of these compounds or their more stable derivatives in treatment of MS.
Sigma-Aldrich
Poly(Ala, Glu, Lys, Tyr) 6:2:5:1 hydrobromide, Ala:Glu:Lys:Tyr (6:2:5:1), mol wt 20,000-30,000