- Preliminary experimental results of gas recycling subsystems except carbon dioxide concentration.
Preliminary experimental results of gas recycling subsystems except carbon dioxide concentration.
Advances in space research : the official journal of the Committee on Space Research (COSPAR) (1987-01-01)
K Otsuji, T Sawada, S Satoh, S Kanda, H Matsumura, S Kondo, K Otsubo
PMID11537272
ABSTRACT
Oxygen concentration and separation is an essential factor for air recycling in a CELSS. Furthermore, if the value of the plant assimilatory quotient is not coincident with that of the animal respiratory quotient, the recovery of O2 from the concentrated CO2 through chemical methods will become necessary to balance the gas contents in a CELSS. Therefore, oxygen concentration and separation equipment using Salcomine and O2 recovery equipment, such as Sabatier and Bosch reactors, were experimentally developed and tested.