MilliporeSigma
  • [The preventing and control of ginseng Rhizoctonia solani with nymexazol].
[The preventing and control of ginseng Rhizoctonia solani with nymexazol].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (1997-02-01)
M Shun, X Lu
ABSTRACT
This paper reports the effect on nymexazol protecting the Rhizoctonia solani of Panax ginseng. The protective effect is in 80-95%. The drug can substitute quintozene and fenaminosulf in production.
Sigma-Aldrich
Fenaminosulf, ≥98%
Supelco
Fenaminosulf, PESTANAL®, analytical standard