MilliporeSigma
  • Fate of MTBE and DCPD compounds relative to BTEX in gasoline-contaminated aquifers.
Fate of MTBE and DCPD compounds relative to BTEX in gasoline-contaminated aquifers.
TheScientificWorldJournal (2003-06-14)
L Olivella, M Figueras, J Fraile, M Vilanova, A Ginebreda, D Barceló
ABSTRACT
The aim of this communication is to provide preliminary results on MTBE monitoring, and at the same time to propose some new tracers of gasoline pollution in groundwater. An overview is presented on benzene-toluene-ethylbenzene-xylene (BTEX), methyl tertiary-butyl ether (MTBE), and dicyclopentadienes (DCPD) contents in gasoline formulations. Their specific fate in gasoline-contaminated aquifers are consistent with their physical-chemical properties.
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mg
Estimated to ship on March 23, 2025
$357.00
$267.75