MilliporeSigma
  • Resistance of medical gloves to permeation by methyl methacrylate (MMA), ethylene glycol dimethacrylate (EGDMA), and 1,4-butanediol dimethacrylate (1,4-BDMA).
Resistance of medical gloves to permeation by methyl methacrylate (MMA), ethylene glycol dimethacrylate (EGDMA), and 1,4-butanediol dimethacrylate (1,4-BDMA).
International journal of occupational safety and ergonomics : JOSE (2003-10-28)
Emma-Christin Lönnroth, I Eystein Ruyter
ABSTRACT
Gloves afford hand protection by minimizing skin contact. The effectiveness of medical gloves to protect against permeation of the monomers, methyl methacrylate (MMA), ethylene glycol dimethacrylate (EGDMA), and 1,4-butanediol dimethacrylate (1,4-BDMA), was assessed focusing on permeation rates and degradation of glove materials caused by monomer contact. Fifteen different brands of gloves were tested using a European Standard procedure. Surface images of glove materials before and after exposure to the monomer mixture were obtained using a scanning electron microscope. The standard is not applicable as the only method for estimating the safety of gloves, but it is useful as guideline together with the cumulative permeation of acrylic monomers. Monomer contact on the outside resulted in substantial swelling of most glove materials, and structure changes of the inside surface.
Sigma-Aldrich
1,4-Butanediol dimethacrylate, 95%, contains 200-300 ppm MEHQ as inhibitor
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
$220.00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
$391.00