Merck
  • Beta-glucosidase, exo-beta-glucanase and pyridoxine transglucosylase activities of rice BGlu1.
Beta-glucosidase, exo-beta-glucanase and pyridoxine transglucosylase activities of rice BGlu1.
The Biochemical journal (2003-12-25)
Rodjana Opassiri, Yanling Hua, Onnop Wara-Aswapati, Takashi Akiyama, Jisnuson Svasti, Asim Esen, James R Ketudat Cairns
ABSTRACT
The bglu1 cDNA for a beta-glucosidase cloned from rice (Oryza sativa L.) seedlings was expressed as a soluble and active protein in Escherichia coli and designated BGlu1. This enzyme hydrolysed beta-1,4-linked oligosaccharides with increasing catalytic efficiency (kcat/Km) values as the DP (degree of polymerization) increased from 2 to 6. In contrast, hydrolysis of beta-1,3-linked oligosaccharides decreased from DP 2 to 3, and polymers with a DP greater than 3 were not hydrolysed. The enzyme also hydrolysed p -nitrophenyl beta-D-glycosides and some natural glucosides but with lower catalytic efficiency than beta-linked oligosaccharides. Pyridoxine 5'-O-beta-D-glucoside was the most efficiently hydrolysed natural glycoside tested. BGlu1 also had high transglucosylation activity towards pyridoxine, producing pyridoxine 5'-O-beta-D-glucopyranoside in the presence of the glucose donor p-nitrophenyl beta-D-glucoside.
Sigma-Aldrich
4-Nitrophenyl β-D-glucopyranoside, ≥98% (TLC)