MilliporeSigma
  • Age-related variation of plasma concentrations of skatole, androstenone, testosterone, oestradiol-17 beta, oestrone sulphate, dehydroepiandrosterone sulphate, triiodothyronine and IGF-1 in six entire male pigs.
Age-related variation of plasma concentrations of skatole, androstenone, testosterone, oestradiol-17 beta, oestrone sulphate, dehydroepiandrosterone sulphate, triiodothyronine and IGF-1 in six entire male pigs.
Reproduction in domestic animals = Zuchthygiene (2004-06-09)
G Zamaratskaia, J Babol, A Madej, E J Squires, K Lundström
ABSTRACT
This study describes the age-related variation in boar taint compounds, skatole and androstenone, and testosterone, oestradiol-17 beta (E17 beta), oestrone sulphate (ES), dehydroepiandrosterone sulphate (DHEAS), triiodothyronine (T(3)) and insulin-like growth factor-1 (IGF-1) in six boars. Three pairs of littermates of crossbred entire male pigs (from three Yorkshire x Duroc dams and one Hampshire sire) were included. Blood samples were taken at the age of 9-15 weeks and thereafter at weekly intervals from the age of 20-32 weeks. Plasma concentrations of skatole, androstenone, testosterone, E17 beta, ES, DHEAS, T(3) and IGF-1 were measured. We found that skatole levels in boars increased at the age around puberty after an increase in the levels of testicular steroids. Levels of skatole were not associated with the levels of sex steroids, T(3) and IGF-1. However, the increased level of testicular steroids is probably the underlying factor needed for high skatole levels to occur although the specific mechanism leading to increased skatole levels remains unknown.
Sigma-Aldrich
5α-Androst-16-en-3-one