Merck
First practical colorimetric assay of lithium in serum.
Clinical chemistry (1992-09-01)
E Chapoteau, B P Czech, W Zazulak, A Kumar
ABSTRACT
We report a colorimetric method for determining lithium in blood serum without sample pretreatment or solvent-extraction steps. The method is based on a novel chromogenic ionophore that exhibits exceptionally high selectivity (much greater than 4000:1) for lithium over sodium. The standard curve for the method is linear up to 3.5 mmol/L and exceeds the therapeutic range for lithium. The results for 57 patients' samples correlated well with results obtained with an Instrumentation Laboratory flame photometer (r = 0.97).
Sigma-Aldrich
Diethylene glycol methyl ether, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethylene glycol methyl ether, ReagentPlus®, ≥99.0%