MilliporeSigma
  • Stereospecific uncatalyzed alpha-O-glycosylation and alpha-C-glycosidation by means of a new D-Gulal-derived alpha vinyl oxirane.
Stereospecific uncatalyzed alpha-O-glycosylation and alpha-C-glycosidation by means of a new D-Gulal-derived alpha vinyl oxirane.
The Journal of organic chemistry (2004-10-09)
Valeria Di Bussolo, Micaela Caselli, Maria Rosaria Romano, Mauro Pineschi, Paolo Crotti
ABSTRACT
The reaction of alpha vinyl oxirane 5, prepared through a new route to the d-gulal system, with O-nucleophiles (alcohols and di-O-isopropylidene-alpha-d-monosaccharides) and C-nucleophiles (lithium alkyls) affords, in a completely stereoselective way, the corresponding 2-unsaturated alpha O- and C-glycosides having the same configuration as the starting epoxide.
Sigma-Aldrich
3,4-Epoxy-1-butene, 98%