MilliporeSigma
  • Facile exchange of glycosyl S,S-acetals to their O,O-acetals and preparation of glycofuranosides from acyclic glycosyl S,S-acetals under metal-free reaction conditions in the presence of 1,3-dibromo-5,5-dimethylhydantoin.
Facile exchange of glycosyl S,S-acetals to their O,O-acetals and preparation of glycofuranosides from acyclic glycosyl S,S-acetals under metal-free reaction conditions in the presence of 1,3-dibromo-5,5-dimethylhydantoin.
Carbohydrate research (2005-02-01)
Soni Kamlesh Madhusudan, Anup Kumar Misra
ABSTRACT
Exchange of acyclic glycosyl dithioacetals to their O,O-acetals has been achieved by a generalized reaction protocol mediated by 1,3-dibromo-5,5-dimethylhydantoin under mild, metal-free and neutral conditions. This methodology has been extended to the synthesis of alkyl glycofuranosides.
Sigma-Aldrich
1,3-Dibromo-5,5-dimethylhydantoin, 98%