MilliporeSigma
  • Synthesis of the beta-hydroxydopa-gamma-hydroxy-delta-sulfinylnorvaline component of ustiloxins A and B.
Synthesis of the beta-hydroxydopa-gamma-hydroxy-delta-sulfinylnorvaline component of ustiloxins A and B.
Organic & biomolecular chemistry (2005-02-26)
Luke Hunter, Malcolm D McLeod, Craig A Hutton
ABSTRACT
The dopa-sulfinylnorvaline component of ustiloxins A and B has been prepared using an Evans salen-Al-catalysed aldol reaction with a 6-mercaptoisovanillin derivative, followed by incorporation of the norvaline group and asymmetric oxidation of the resulting sulfide to give the corresponding sulfoxide.
Sigma-Aldrich
3-Hydroxy-4-methoxybenzaldehyde, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
L-Norvaline, arginase inhibitor
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
DL-Norvaline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00
Sigma-Aldrich
Isovanillin, ≥95.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Estimated to ship on March 23, 2025
$161.00