Merck
  • Effects of purified herbal extract of Salvia miltiorrhiza on ischemic rat myocardium after acute myocardial infarction.
Effects of purified herbal extract of Salvia miltiorrhiza on ischemic rat myocardium after acute myocardial infarction.
Life sciences (2005-04-06)
Jian Sun, Shan Hong Huang, Benny K-H Tan, Matt Whiteman, Yi Chun Zhu, Ya Jun Wu, Yeekong Ng, Wei Duan, Yi Zhun Zhu
ABSTRACT
In the current study, we compared purified Salvia miltiorrhiza extract (PSME) with Angiotensin-converting enzyme inhibitor, Ramipril, in in vitro experiments and also in vivo using animal model of myocardial infarction. PSME was found to have a significantly higher trolox equivalent antioxidant capacity which indicated a great capacity for scavenging free radicals. PSME could also prevent pyrogallo red bleaching and DNA damage. After 2 weeks treatment with PSME or Ramipril, survival rates of rats with experimental myocardial infarction were marginally increased (68.2% and 71.4%) compared with saline (61.5%). The ratios of infarct size to left ventricular size in both PSME-and Ramipril-treated rats were significantly less than that in the saline-treated group. Activity of cardiac antioxidant enzyme superoxide dismutase (SOD) was significant higher while level of Thiobarbituric acid-reactive substances (TBARs) was lower in the PSME treated group. Purified and standardized Chinese herb could provide an alternative regimen for the prevention of ischemic heart disease.
Sigma-Aldrich
Pyrogallol Red, Suitable for use as a complexometric indicator