MilliporeSigma
Alpha-tocopheryl phosphate: a novel, natural form of vitamin E.
Free radical biology & medicine (2005-09-06)
Robert Gianello, Roksan Libinaki, Angelo Azzi, Paul D Gavin, Yesim Negis, Jean-Marc Zingg, Phillip Holt, Hooi-Hong Keah, Annike Griffey, Andrew Smallridge, Simon M West, Esra Ogru
ABSTRACT
We have detected alpha-tocopheryl phosphate in biological tissues including liver and adipose tissue, as well as in a variety of foods, suggesting a ubiquitous presence in animal and plant tissue. Alpha-tocopheryl phosphate is a water-soluble molecule that is resistant to both acid and alkaline hydrolysis, making it undetectable using standard assays for vitamin E. A new method was therefore developed to allow the extraction of both alpha-tocopheryl phosphate and alpha-tocopherol from a single specimen. We used ESMS to detect endogenous alpha-tocopheryl phosphate in biological samples that also contained alpha-tocopherol. Due to the significance of these findings, further proof was required to unequivocally demonstrate the presence of endogenous alpha-tocopheryl phosphate in biological samples. Four independent methods of analysis were examined: HPLC, LCMS, LCMS/MS, and GCMS. Alpha-tocopherol phosphate was identified in all instances by comparison between standard alpha-tocopheryl phosphate and extracts of biological tissues. The results show that alpha-tocopheryl phosphate is a natural form of vitamin E. The discovery of endogenous alpha-tocopheryl phosphate has implications for the expanding knowledge of the roles of alpha-tocopherol in biological systems.
Sigma-Aldrich
(±)-α-Tocopherol phosphate disodium salt, water-soluble α-tocopherol analog