MilliporeSigma
  • Copper-catalyzed arylation of amines using diphenyl pyrrolidine-2-phosphonate as the new ligand.
Copper-catalyzed arylation of amines using diphenyl pyrrolidine-2-phosphonate as the new ligand.
The Journal of organic chemistry (2005-11-10)
Honghua Rao, Hua Fu, Yuyang Jiang, Yufen Zhao
ABSTRACT
[Chemical reaction: See text] We have developed a general, efficient, and inexpensive catalyst system for arylation of amines by using 10 mol % of CuI as the copper source, 20 mol % of diphenyl pyrrolidine-2-phosphonate (DPP) as the ligand, K3PO4 as the base, and DMF containing 2% water (v/v) as the solvent.
Sigma-Aldrich
1-Phenyldodecane, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.2 mL
In Stock
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1537
$530.00
Supelco
1-Phenyldodecane, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.2 mL
In Stock
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1537
$530.00