Merck
  • Potential impact of steatosis on cytochrome P450 enzymes of human hepatocytes isolated from fatty liver grafts.
Potential impact of steatosis on cytochrome P450 enzymes of human hepatocytes isolated from fatty liver grafts.
Drug metabolism and disposition: the biological fate of chemicals (2006-06-10)
M Teresa Donato, Agustín Lahoz, Nuria Jiménez, Gabriela Pérez, Alfonso Serralta, José Mir, José V Castell, M José Gómez-Lechón
ABSTRACT
Liver grafts discarded for transplantation because of macrosteatosis can constitute a valuable source of human hepatocytes for in vitro metabolic and pharmacotoxicological studies or for therapeutic applications. A condition for using hepatocyte suspensions for these purposes is the preservation of their metabolic competence and, particularly, drug-metabolizing enzymes. A reduction in microsomal cytochrome P450 (P450) activities was observed in fatty livers (>40% steatosis) with respect to normal tissue. Similarly, decreased levels of 7-ethoxycoumarin O-deethylation and testosterone metabolism were observed in human hepatocyte cultures prepared from steatotic liver tissue. To clarify the potential impact of lipid accumulation on human hepatic P450 enzymes, we have used an in vitro model of "cellular steatosis" by incubation of cultured hepatocytes with increasing concentrations (0.25-3 mM) of long-chain free fatty acids (FFA). A dose-dependent accumulation of lipids in the cytosol is induced by FFA mixture. Hepatocytes exposed to 1 mM FFA for 14 h showed lower activity values of CYP1A2, CYP2A6, CYP2B6, CYP2C9, CYP2D6, CYP2E1, and CYP3A4 enzymes than nontreated hepatocytes (about 45-65% reduction). This treatment also produced significant decreases in CYP1A2, CYP2A6, CYP2C9, CYP2D6, CYP2E1, and CYP3A4 mRNA to about 55 to 75% of mRNA levels in control cells. Our results suggest that although human hepatocytes isolated from steatotic liver show reduced P450 activities, they are metabolically competent and can be used for drug metabolism studies.
Sigma-Aldrich
Resorufin methyl ether