Merck
  • Paracoccus bengalensis sp. nov., a novel sulfur-oxidizing chemolithoautotroph from the rhizospheric soil of an Indian tropical leguminous plant.
Paracoccus bengalensis sp. nov., a novel sulfur-oxidizing chemolithoautotroph from the rhizospheric soil of an Indian tropical leguminous plant.
Systematic and applied microbiology (2006-07-11)
Wriddhiman Ghosh, Sukhendu Mandal, Pradosh Roy
ABSTRACT
Paracoccus versutus-like isolates from the rhizosphere of Clitoria ternatea, a slender leguminous herb (family--Papilionaceae), found ubiquitously in waste places and village forests of the Lower Gangetic plains of India, presented a case of graduated infraspecific variation that was capped by the identification of a new species Paracoccus bengalensis (type strain JJJ(T) = LMG 22700(T) = MTCC 7003(T)). The diverged phenetic and genetic structure of these sulfur-oxidizing chemolithoautotrophs presented a case of apparent nonconformity of 16S rRNA gene sequence similarities with results of DNA-DNA hybridization. Despite high 16S rRNA gene sequence similarity with P. versutus one of the newly isolated strains, viz., JJJ(T) was identified as a new species of Paracoccus by virtue of its explicitly low DNA-DNA hybridization (42-45%) with the type strain of the closest species P. versutus (), distinct G + C content (65.3 mol%), physiological and biochemical differences amounting to <60% phenetic similarity with strains of P. versutus as well as new isolates akin to the species. The newly described species also had a unique fatty acid profile that was distinguished by the absence of 18:1 omega9c, unique possession of Summed feature 3 (16:1omega7c & 15:0 iso 2-OH), 19:0 10 methyl, and a much higher concentration of 19:0 cycloomega8c.
Sigma-Aldrich
Sodium tetrathionate dihydrate, ≥98% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium tetrathionate dihydrate, purum p.a., ≥98.0% (T)