MilliporeSigma
  • The alarm reaction of coho salmon parr is impaired by the carbamate fungicide IPBC.
The alarm reaction of coho salmon parr is impaired by the carbamate fungicide IPBC.
Aquatic toxicology (Amsterdam, Netherlands) (2006-07-21)
Keith B Tierney, Amber L Taylor, Peter S Ross, Christopher J Kennedy
ABSTRACT
To determine whether the carbamate fungicide IPBC alters the olfactory-mediated behavioral and physiologic alarm responses of coho salmon parr (Oncorhynchus kisutch), groups of coho were exposed to skin extract (an alarm pheromone source) under a variety of conditions. In the 3min following skin extract exposure, freezing behavior was significantly increased (In the 3 min following skin extract exposure, freezing behavior was significantly increased under darkness (IR lighting) but not ambient lighting (25.3+/-2.6% and 7.5+/-5.7%, respectively; Delta calculated as: [(time (s) after/time (s) before)-1]x100%), and so IR was used for further experiments. Physiologically, following skin extract exposure, plasma cortisol concentration was increased at 0.5h (58.1+/-14.6ng/ml versus 4.32+/-1.31ng/ml, exposed versus control), hematocrit (Hct) was increased at 2h (50.4+/-1.0% versus 41.7+/-1.6%), and leucocrit (Lct) was decreased at 0.5 and 2h (0.534+/-0.114 and 0.13+/-0.01% versus 1.23+/-0.20%). After 0.5h exposures to 0, 1, 10 and 100microg/l IPBC and skin extract, the time spent dashing (>5cm/s) increased significantly (323+/-118%) in the first minute after skin extract exposure, but was absent in IPBC-exposed coho. Freezing behavior increased after skin extract exposure with control and 1microg/l IPBC exposures (11.0+/-3.0% and 17.7+/-11.0%, respectively), but was absent after 10microg/l and decreased after 100microg/l IPBC. Physiologically, Hct and plasma lactate concentration were significantly increased above controls after 1microg/l IPBC exposure (Hct: 45.7+/-1.6% versus 34.0+/-1.6%, lactate: 12.8+/-1.2mM versus 3.30+/-1.2mM). After 10microg/l exposure, IPBC alone elicited a stress response similar to skin extract. However in the 100microg/l treatment group the stress parameters were not different from controls. These findings suggest that the behavioral and physiologic alarm responses of juvenile salmonids may be impaired by acute exposure to > or =1microg/l IPBC.
Sigma-Aldrich
3-Iodo-2-propynyl N-butylcarbamate, 97%