MilliporeSigma
  • Vibrational assignment and structure of trifluorobenzoylacetone. A density functional theoretical study.
Vibrational assignment and structure of trifluorobenzoylacetone. A density functional theoretical study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-07-25)
Sayyed Faramarz Tayyari, Mohammad Vakili, Abdo-Reza Nekoei, Hedayat Rahemi, Yan Alexander Wang
ABSTRACT
Molecular structure and vibrational frequencies of 4,4,4-trifluoro-1-phenyl-1,3-butanedione, known as trifluorobenzoylacetone (TFBA), have been investigated by means of density functional theory (DFT) calculations. The results were compared with those of benzoylacetone (BA), acetylacetone (AA), and trifluoroacetylacetone (TFAA). Comparing the calculated and experimental band frequencies and intensities suggests coexisting of both stable cis-enol conformers in comparable proportions in the sample. The energy difference between the two stable chelated enol forms is negligible, 0.96 kcal/mol, calculated at B3LYP/6-311++G** level of theory. The molecular stability and the hydrogen bond strength were investigated by applying the natural bond orbital (NBO) theory and geometry calculations. The theoretical calculations and spectroscopic results indicate that the hydrogen bond strength of TFBA is between those of TFAA and AA, considerably weaker than that of BA.
Sigma-Aldrich
1-Phenyl-1,3-butanedione, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on February 17, 2025
Details...
₩122,612