Merck
  • Basic investigations on the performance of a normoxic polymer gel with tetrakis-hydroxy-methyl-phosphonium chloride as an oxygen scavenger: reproducibility, accuracy, stability, and dose rate dependence.
Basic investigations on the performance of a normoxic polymer gel with tetrakis-hydroxy-methyl-phosphonium chloride as an oxygen scavenger: reproducibility, accuracy, stability, and dose rate dependence.
Medical physics (2006-08-11)
Christian Bayreder, Dietmar Georg, Ewald Moser, Andreas Berg
ABSTRACT
Magnetic resonance (MR)-based polymer gel dosimetry using normoxic polymer gels, represents a new dosimetric method specially suited for high-resolution three-dimensional dosimetric problems. The aim of this study was to investigate the dose response with regard to stability, accuracy, reproducibility, and the dose rate dependence. Tetrakis-hydroxy-methyl-phosphonium chloride (THPC) is used as an oxygen scavenger, and methacrylic acid as a monomer. Accuracy, reproducibility, and dose resolution were determined for MR protocols at low spatial resolution (typical for clinical scanners), medium, and microimaging-resolution protocols at three different dose levels. The dose-response stability and preirradiation-induced variations in R2, related to the time interval between preparation and irradiation of the polymer gel, were investigated. Also postirradiation stability of the polymer gel was considered. These experiments were performed using a 60Co beam (E = 1.2 MV) in a water phantom. Moreover, we investigated the dose rate dependence in the low, medium, and saturation dose region of the normoxic polymer gel using a linear accelerator at photon energy of 25 MV. MR scanning was performed on a 3 T whole body scanner (MEDSPEC 30/80, BRUKER BIOSPIN, Ettlingen, Germany) using several coils and different gradient systems adapted to the acquired spatial resolution investigated. For T2-parameter selective imaging and determination of the relaxation rate R2 = 1/T2, a multiple spin echo sequence with 20 equidistant echoes was used. With regard to preirradiation induced variations R2 increases significantly with the increasing time interval between the polymer gel preparation and irradiation. Only a slight increase in R2 can be observed for varying the postirradiation-time solely. The dose reproducibility at voxel volumes of about 1.4 x 1.4 x 2 mm3 is better than 2%. The accuracy strongly depends on the calibration curve. THPC represents a very effective oxygen scavenger in methacrylic acid and gelatin. Polymer gels containing THPC offer high sensitivity to dose but their dose response also strongly depends on dose rate in the medium and high dose region.
Sigma-Aldrich
Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution, technical, 70-75% in H2O (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetrakis(hydroxymethyl)phosphonium chloride solution, 80% in H2O