Merck
  • Multiple-element detection in aqueous solution and seawater by using an on-line preconcentration method for inductively coupled plasma mass spectrometry.
Multiple-element detection in aqueous solution and seawater by using an on-line preconcentration method for inductively coupled plasma mass spectrometry.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-10-14)
Li-Shing Huang, King-Chuen Lin
ABSTRACT
Inductively coupled plasma-mass spectrometry (ICP-MS) incorporated with an on-line preconcentration system was used to determine trace amounts of ten metals including Ni, Cu, Zn, Rh, Ag, Cd, In, Au, Tl, and Pb in aqueous solutions and seawater. These metals, which formed the complexes, were retained in a sorbent microcolumn, followed by elution with methanol through a desolvation unit, which was capable of removing 83% of methanol. The limits of detection for these elements were determined simultaneously to be in the range from 3 to 20 ng/L.
Sigma-Aldrich
Diethyl dithiophosphate ammonium salt, 95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
O,O′-Diethyl dithiophosphate, technical grade, 90%