MilliporeSigma
  • Hemoglobin protein hollow shells fabricated through covalent layer-by-layer technique.
Hemoglobin protein hollow shells fabricated through covalent layer-by-layer technique.
Biochemical and biophysical research communications (2007-01-24)
Li Duan, Qiang He, Xuehai Yan, Yue Cui, Kewei Wang, Junbai Li
ABSTRACT
Hemoglobin (Hb) protein microcapsules held together by cross-linker, glutaraldehyde (GA), were successfully fabricated by covalent layer-by-layer (LbL) technique. The Schiff base reaction occurred on the colloid templates between the aldehyde groups of GA and free amino sites of Hb results in the formation of GA/Hb microcapsules after the removal of the templates. The structure of obtained monodisperse protein microcapsule was characterized by transmission electron microscopy (TEM) and confocal laser scanning microscopy (CLSM). The UV-Vis spectra measurements demonstrate the existence of Hb in the assembled capsules. Cyclic voltammetry (CV) and potential-controlled amperometric measurements (I-t curve) confirm that hemoglobin microcapsules after fabrication remain their heme electroactivity. Moreover, direct electron transfer process from protein to electrode surface was performed to detect the heme electrochemistry without using any mediator or promoter. The experiments of fluorescence recovery after photobleaching (FRAP) by CLSM demonstrate that the hemoglobin protein microcapsules have an improved permeability comparing to the conventional polyelectrolyte microcapsules.
Sigma-Aldrich
Manganese(II) carbonate, ≥99.9% trace metals basis