MilliporeSigma
  • A novel approach to cyclin-dependent kinase 5/p25 inhibitors: A potential treatment for Alzheimer's disease.
A novel approach to cyclin-dependent kinase 5/p25 inhibitors: A potential treatment for Alzheimer's disease.
Bioorganic & medicinal chemistry (2007-07-24)
Mahendra Ramesh Shiradkar, Mallikarjuna Bandrehally Padhalingappa, Sastry Bhetalabhotala, Kalyan Chakravarthy Akula, Dattu Anna Tupe, Raghotham Reddy Pinninti, Suman Thummanagoti
ABSTRACT
Based on the earlier results of our in-house database and compound library, a series of novel clubbed thienyl triazoles was designed which may emerge as potential cdk5/p25 inhibitors, for the treatment of Alzheimer's disease. A benign synthesis was planned so as to take an advantage of MAOS (Microwave Assisted Organic Synthesis) method. Evaluation of the SAR of this series has allowed the identification of compounds 4, 5, 7 and 8 from series I while 13, 14, 16 and 17 from series II as significant cdk5/p25 inhibitors and thus have potential as possible treatments for Alzheimer's disease.
Sigma-Aldrich
Tienilic Acid, ≥98% (HPLC)