MilliporeSigma
  • Aqueous self-assembly of phytantriol in ternary systems: effect of monoolein, distearoylphosphatidylglycerol and three water-miscible solvents.
Aqueous self-assembly of phytantriol in ternary systems: effect of monoolein, distearoylphosphatidylglycerol and three water-miscible solvents.
Journal of colloid and interface science (2007-07-28)
Pia Wadsten-Hindrichsen, Johanna Bender, Johan Unga, Sven Engström
ABSTRACT
The aqueous phase behavior of phytantriol (PT) in mixtures of monoolein (MO), distearoylphosphatidylglycerol (DSPG), propylene glycol (PG), polyethylene glycol 400 (PEG 400) and 2-methyl-2,4-pentanediol (MPD) was investigated by visual inspection, polarized light microscopy and small angle X-ray diffraction at room temperature. The phase diagrams of PT and MO in water are qualitatively very similar and PT/MO mixtures in excess water form one cubic phase of space group Pn3m irrespective of mixing ratio. The addition of the charged membrane lipid DSPG to the PT system gives rise to a considerable water swelling of the cubic phases as well as the occurrence of a cubic phase of space group Im3m. Whereas all three solvents studied give rise to a sponge (L3) phase in the MO-water system, this phase was only found when MPD was added to the PT-water system. The results are discussed with respect to the chemical differences between PT and MO.
Sigma-Aldrich
Hexylene glycol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Hexylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Hexylene glycol, puriss., ≥99.0% (GC)
Sigma-Aldrich
Hexylene glycol, 99%