Merck
  • [Toxicity of 4-chloro-2-nitroaniline and 2-chloro-4-nitroaniline to isolated rat hepatocytes].
[Toxicity of 4-chloro-2-nitroaniline and 2-chloro-4-nitroaniline to isolated rat hepatocytes].
La Medicina del lavoro (1991-05-01)
D Cottalasso, M A Pronzato, C Domenicotti, G Barisione, L Fontana, G Nanni
ABSTRACT
The toxicity of 4-chloro-2-nitroaniline (4C2NA) and 2-chloro-4-nitroaniline (2C4NA) was investigated on isolated rat hepatocytes following 1-3 hours of exposure to 0.2 and/or 2 mM of these xenobiotics. The higher of the two concentrations appeared to induce a statistically significant loss of cellular viability (p less than 0.01 compared to control), judged by Trypan Blue staining, after 3 hours of incubation with these substances means = 58, SD = 7%; and means = SD = 7%; for 4C2NA and 2C4NA, respectively). Furthermore, both chloronitroanilines produced an hepatocellular and microsomal damage demonstrated by conspicuous changes in LDH and G-6-Pase activities (p less than 0.01). The exposure to 2 mM of both 4C2NA and/or 2C4NA produced a marked depletion of the intracellular pool of GSH after 3 hours (13 mM/10(6) and 10 mM/10(6) cells, respectively; p less than 0.01). Thus it seems possible that 2C4NA may induce a more severe cellular damage than that induced by 4C2NA.
Sigma-Aldrich
2-Chloro-4-nitroaniline, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Chloro-2-nitroaniline, 99%