Merck
  • [Potential use of the herbicide quizalofop-p-ethyl for eicosapentaenoic acid overproduction by the diatom Nitzschia laevis].
[Potential use of the herbicide quizalofop-p-ethyl for eicosapentaenoic acid overproduction by the diatom Nitzschia laevis].
Sheng wu gong cheng xue bao = Chinese journal of biotechnology (2007-12-07)
Xiao-Hong Cao, Song-Yao Li, Chun-Ling Wang, Mei-Fang Lu
ABSTRACT
The diatom Nitzschia laevis is a good alternative source of eicosapentaenoic acid (EPA). Besides strategies for high cell density culture, EPA productivity may be further improved by herbicides. The effect of the herbicide quizalofop-p-ethyl on the growth and EPA production was studied in this paper. As the solvent of the herbicide, DMSO was proved to inhibit the growth and EPA production of N. laevis. The concentration of DMSO in the medium should not exceed 0.2%. Quizalofop-p-ethyl could cause morphology damage to the N. laevis cells. With the increasing concentration of quizalofop-p-ethyl from 0 mmol/L to 0.4 mmol/L, the dry cell weight production decreased, while at the same time, the lipid content of the dry cell mass increased. When treated with 0.1 mmol/L quizalofop-p-ethyl, the EPA content increased from 3.00% to 3.58% (of dry cell weight, DW), and the proportion of EPA (20:5) in total fatty acids (TFA) increased from 25.15% to 32.88% . These results indicated that the herbicide quizalofop-p-ethyl could stimulate the accumulation of EPA; therefore it might be useful for selecting algae colonies that overproduce EPA.
Product Description
Supelco
Quizalofop-P-ethyl, PESTANAL®, analytical standard
Supelco
Quizalofop-ethyl, PESTANAL®, analytical standard