MilliporeSigma
Retinoic acid induced suicidal erythrocyte death.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2008-01-23)
Olivier M Niemoeller, Michael Foller, Camelia Lang, Stephan M Huber, Florian Lang
ABSTRACT
Vitamin A and retinoic acid have previously been shown to confer some protection against a severe course of malaria by fostering the phagocytosis of parasitized erythrocytes. Phagocytosis of erythrocytes is stimulated by phosphatidylserine exposure at the cell surface. The present study has thus been performed to explore the effect of retinoic acid and the specific retinoic acid receptor (RAR) agonist 4-(E-2-[5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl]-1-propenyl) benzoic acid (TTNPB) on erythrocyte annexin V binding, which reflects phosphatidylserine exposure at the cell surface. A 24 hours exposure to either, retinoic acid (3 microM) or TTNPB (3 microM), indeed significantly increased annexin binding, an effect paralleled by decrease of forward scatter reflecting cell shrinkage. According to Fluo3 fluorescence, exposure to either, retinoic acid (10 microM, 24 hours) or TTNPB (10 microM, 6 hours), significantly increased cytosolic Ca(2+)-activity, a known trigger of phosphatidylserine exposure. Infection of erythrocytes with Plasmodium falciparum increased phosphatidylserine exposure, an effect increased in the presence of TTNPB. In conclusion, retinoid acid and TTNPB trigger phosphatididylserine exposure and cell shrinkage of erythrocytes, typical features of suicidal erythrocyte death or eryptosis. The eryptosis could participate in the accelerated clearance of parasitized erythrocytes from circulating blood following treatment with retinoids.
Sigma-Aldrich
TTNPB