MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • de novo design and synthesis of N-benzylanilines as new candidates for VEGFR tyrosine kinase inhibitors.
de novo design and synthesis of N-benzylanilines as new candidates for VEGFR tyrosine kinase inhibitors.
Organic & biomolecular chemistry (2008-03-11)
Masaharu Uno, Hyun Seung Ban, Wataru Nabeyama, Hiroyuki Nakamura
ABSTRACT
N-Benzylanilines were designed and synthesized as vascular endothelial growth factor (VEGF)-2 inhibitors using de novo drug design systems based on the X-ray structure of VEGFR-2 kinase domain. Among compounds synthesized, compound showed the most potent inhibitory activity toward VEGFR-2 (KDR) tyrosine kinase and its IC(50) value was 0.57 microM.
Sigma-Aldrich
N-Benzylaniline, ≥99%