MilliporeSigma
  • Ultrasmall c(RGDyK)-coated Fe3O4 nanoparticles and their specific targeting to integrin alpha(v)beta3-rich tumor cells.
Ultrasmall c(RGDyK)-coated Fe3O4 nanoparticles and their specific targeting to integrin alpha(v)beta3-rich tumor cells.
Journal of the American Chemical Society (2008-05-27)
Jin Xie, Kai Chen, Ha-Young Lee, Chenjie Xu, Andrew R Hsu, Sheng Peng, Xiaoyuan Chen, Shouheng Sun
ABSTRACT
We report a direct synthesis of ultrasmall c(RGDyK) peptide-coated Fe3O4 NPs (<10 nm in hydrodynamic diameter) and demonstrate their in vivo tumor-specific targeting capability. The Fe3O4 NPs are synthesized by thermal decomposition of iron pentacarbonyl in the presence of 4-methylcatechol (4-MC), and the peptide is coupled to the nanoparticles through 4-MC via Mannich reaction. The c(RGDyK)-MC-Fe3O4 NPs have an overall diameter of approximately 8.4 nm and are stable in physiological conditions. When administrated intravenously, these c(RGDyK)-MC-Fe3O4 NPs accumulate preferentially in the integrin alphavbeta3-rich tumor area, which are readily tracked by MRI.
Sigma-Aldrich
4-Methylcatechol, ≥95%