MilliporeSigma
  • Lewis base catalyzed 1,3-dithiane addition to carbonyl and imino compounds using 2-trimethylsilyl-1,3-dithiane.
Lewis base catalyzed 1,3-dithiane addition to carbonyl and imino compounds using 2-trimethylsilyl-1,3-dithiane.
Chemistry, an Asian journal (2008-06-21)
Makoto Michida, Teruaki Mukaiyama
ABSTRACT
Lewis base-catalyzed 1,3-dithiane addition to electrophiles such as carbonyl compounds and N-substituted aldimines with 2-trimethylsilyl-1,3-dithiane (TMS-dithiane) is described. By the activation of the carbon-silicon bond in the presence of a Lewis base catalyst such as tetrabutylammonium phenoxide (PhONnBu(4)), a 1,3-dithiane addition reaction proceeded smoothly to afford the corresponding adducts in good to high yields under mild conditions. This synthesis is also applied to the reactions of ketones having alpha-protons, and of N-substituted aldimines.
Sigma-Aldrich
1,3-Dithiane, 97%