MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Measurement of cross sections for the 147Sm(n, alpha)144Nd reaction at 5.0 and 6.0 MeV.
Measurement of cross sections for the 147Sm(n, alpha)144Nd reaction at 5.0 and 6.0 MeV.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2008-08-19)
Guohui Zhang, Jiaguo Zhang, Li'an Guo, Hao Wu, Jinxiang Chen, Guoyou Tang, Yu M Gledenov, M V Sedysheva, G Khuukhenkhuu, P J Szalanski
ABSTRACT
Cross sections of the (147)Sm(n, alpha)(144)Nd reaction were measured at En=5.0 and 6.0MeV. A twin gridded ionization chamber was used as a charged particle detector and two large area (147)Sm(2)O(3) samples placed back to back were employed. Experiments were performed at the 4.5MV Van de Graaff accelerator of Peking University. Neutrons were produced through the D(d, n)(3)He reaction with a deuterium gas target. Absolute neutron flux was determined by a small (238)U fission chamber. Present cross-section data are compared with existing results of evaluations and measurements.
Sigma-Aldrich
Samarium(III) oxide, 99.9% trace metals basis