MilliporeSigma
  • Development and validation of a solid-phase extraction method coupled to liquid chromatography with fluorescence detection for the determination of fluoroquinolone residues in powdered infant formulae. Application to the analysis of samples from the Spanish and Latin American market.
Development and validation of a solid-phase extraction method coupled to liquid chromatography with fluorescence detection for the determination of fluoroquinolone residues in powdered infant formulae. Application to the analysis of samples from the Spanish and Latin American market.
Journal of chromatography. A (2008-10-03)
E Rodriguez, M C Moreno-Bondi, M D Marazuela
ABSTRACT
This paper describes a new method for the effective extraction, clean-up and chromatographic analysis of residues of four fluoroquinolones (ciprofloxacin, enrofloxacin, danofloxacin and sarafloxacin) in powdered infant formulae and follow-on preparations. Samples were reconstituted following the manufacturer's recommendations and treated with trichloroacetic acid in methanol 10% (w/v) for deproteinization. Two solid-phase extraction cartridges have been evaluated for sample clean-up and preconcentration, Strata Screen A and Strata X and the later provided the best recoveries for all the analytes tested. Chromatographic analysis has been carried out using a polar endcapped column (AQUA C(18)) and fluorescence detection, with lomefloxacin (LOME) as internal standard. Method validation has been performed according to European Commission Decision 2002/657/EC criteria, in terms of linearity, recovery, precision, specificity, decision limit (CC(alpha)) and detection capability (CC(beta)). Typical recoveries ranged between 70 and 110% at levels below and above the maximum residue limits of the target analytes in bovine milk, with an excellent intralab reproducibility (RSDs<7%). Matrix effects did not significantly affect method accuracy, as evidenced by analyzing different brands of milk. The method has been successfully applied to the analysis of 100 samples of infant and follow-on formulae of the Spanish and Latin American market, using LC-MS/MS as confirmatory technique.
Product Description
Supelco
Sarafloxacin hydrochloride hydrate, VETRANAL®, analytical standard