MilliporeSigma
  • Development of an enzyme-linked immunosorbent assay for the alpha-cyano pyrethroids multiresidue in Tai lake water.
Development of an enzyme-linked immunosorbent assay for the alpha-cyano pyrethroids multiresidue in Tai lake water.
Journal of agricultural and food chemistry (2009-04-17)
X L Hao, H Kuang, Y L Li, Y Yuan, C F Peng, W Chen, L B Wang, C L Xu
ABSTRACT
A general and broad class selective competitive enzyme-linked immunosorbent assay (ELISA) for the detection of the alpha-cyano pyrethroids was developed. One class specific hapten, the 5-(3-benzylphenyl)-5-cyanopentanoic acid (CPBA-BE), was conjugated with BSA as an immunogen. Polyclonal antibodies, PY-antibodies, were generated. The assay with the most selectivity for the family alpha-cyano pyrethroids such as cyphenothrin was further optimized and tested for tolerance to cosolvent, pH, and ionic strength changes. The IC(50) values of the optimized immunoassay were 4.58 microg L(-1) for cyphenothrin, 5.62 microg L(-1) for fenpropathrin, 7.08 microg L(-1) for deltamethrin, 10.72 microg L(-1) for cypermethrin, 19.95 microg L(-1) for flucythrinate, and 28.18 microg L(-1) for esfenvalerate. The cross-reactivities of cyphenothrin, fenpropathrin, deltamethrin, cypermethrin, flucythrinate, and esfenvalerate were 100%, 81.49%, 64.68%, 42.72%, 22.96%, and 16.25%, respectively. The low detection limit (LDL) of this assay was 0.107 microg L(-1) for cyphenothrin. This method was used for the detection of the alpha-cyano pyrethroids in Tai lake water.
Product Description
Supelco
Cyphenothrin, PESTANAL®, analytical standard, sum of isomers