Merck
  • Evaluation of the toxicity of stress-related aldehydes to photosynthesis in chloroplasts.
Evaluation of the toxicity of stress-related aldehydes to photosynthesis in chloroplasts.
Planta (2009-07-07)
Jun'ichi Mano, Fumitaka Miyatake, Eiji Hiraoka, Masahiro Tamoi
ABSTRACT
Aldehydes produced under various environmental stresses can cause cellular injury in plants, but their toxicology in photosynthesis has been scarcely investigated. We here evaluated their effects on photosynthetic reactions in chloroplasts isolated from Spinacia oleracea L. leaves. Aldehydes that are known to stem from lipid peroxides inactivated the CO(2) photoreduction to various extents, while their corresponding alcohols and carboxylic acids did not affect photosynthesis. alpha,beta-Unsaturated aldehydes (2-alkenals) showed greater inactivation than the saturated aliphatic aldehydes. The oxygenated short aldehydes malondialdehyde, methylglyoxal, glycolaldehyde and glyceraldehyde showed only weak toxicity to photosynthesis. Among tested 2-alkenals, 2-propenal (acrolein) was the most toxic, and then followed 4-hydroxy-(E)-2-nonenal and (E)-2-hexenal. While the CO(2)-photoreduction was inactivated, envelope intactness and photosynthetic electron transport activity (H(2)O --> ferredoxin) were only slightly affected. In the acrolein-treated chloroplasts, the Calvin cycle enzymes phosphoribulokinase, glyceraldehyde-3-phosphate dehydrogenase, fructose-1,6-bisphophatase, sedoheptulose-1,7-bisphosphatase, aldolase, and Rubisco were irreversibly inactivated. Acrolein treatment caused a rapid drop of the glutathione pool, prior to the inactivation of photosynthesis. GSH exogenously added to chloroplasts suppressed the acrolein-induced inactivation of photosynthesis, but ascorbic acid did not show such a protective effect. Thus, lipid peroxide-derived 2-alkenals can inhibit photosynthesis by depleting GSH in chloroplasts and then inactivating multiple enzymes in the Calvin cycle.
Sigma-Aldrich
trans-2-Nonenal, 97%