MilliporeSigma
  • Temperature dependence of the (1)J((11)B(19)F) spin-spin coupling in BF(3) molecule.
Temperature dependence of the (1)J((11)B(19)F) spin-spin coupling in BF(3) molecule.
Magnetic resonance in chemistry : MRC (2009-07-29)
Karol Jackowski, Włodzimierz Makulski, Anna Szyprowska, Andrej Antusek, Michał Jaszuński
ABSTRACT
The (1)J((11)B(19)F) spin-spin coupling of gaseous BF(3) was observed in (11)B NMR spectra as a function of density in a wide range of temperatures. Following the extrapolation of the measured values to the zero-density limit, the coupling constant free from intermolecular effects (1)J(0)((11)B(19)F) was obtained for each temperature. In contrast to previous investigations, the final results indicate a nonlinear dependence of (1)J(0)((11)B(19)F) on temperature. In the corresponding ab initio calculations of spin-spin coupling constants performed at the coupled cluster singles and doubles (CCSD) level to obtain a reliable result for this coupling constant we had to take into account large vibrational corrections.
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Supelco
Boron trifluoride - 1-butanol solution, ~10% in 1-butanol (∼1.3 M), for GC derivatization, LiChropur