MilliporeSigma
Self-trapped vibrational states in synthetic beta-sheet helices.
Chemical communications (Cambridge, England) (2009-07-31)
Erik Schwartz, Pavol Bodis, Matthieu Koepf, Jeroen J L M Cornelissen, Alan E Rowan, Sander Woutersen, Roeland J M Nolte
ABSTRACT
Femtosecond vibrational pump-probe spectroscopy on beta-helical polyisocyanopeptides reveals vibrational self-trapping in the well-defined hydrogen-bonded side groups that is absent when non-hydrogen bonded monomers are mixed in.
Sigma-Aldrich
D-Alanine methyl ester hydrochloride, 98%
Sigma-Aldrich
L-Alanine methyl ester hydrochloride, 99%