MilliporeSigma
Influence of growth conditions on plasmid DNA production.
Journal of microbiology and biotechnology (2009-12-10)
Filomena Silva, Luís Passarinha, Fani Sousa, João A Queiroz, Fernanda C Domingues
ABSTRACT
The obtention of high yields of purified plasmid DNA is viewed as an essential issue to be considered towards efficient production of DNA vaccines and therapeutic plasmids. In this work, Escherichia coli DH5alpha bearing the pVAX1-LacZ plasmid was grown in a developed semi-defined medium at different temperatures and tryptone concentrations. Analysis of pDNA yields and E. coli morphology revealed that at higher temperatures (37 and 40 degrees C), higher specific yields and E. coli filamentation were obtained. However, the best results were achieved when a lower tryptone concentration was used. This approach was shown to be a powerful tool to promote plasmid amplification, keeping the desirable plasmid structure, and favoring the attainment of quality. Our results suggest that by using tryptone alone as an amino acid source, pDNA amplification was improved and a specific yield of 20.43 mg pDNA/g dcw was achieved, proving that this strategy can improve pDNA yield even at a small scale.
Product Description
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from milk solids, Refined hydrolysate
Millipore
Peptone from casein and other animal proteins, suitable for microbiology
Sigma-Aldrich
Peptone from animal tissue, SAJ first grade
Millipore
Peptone from animal tissue, from meat
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture
Millipore
Peptone Primatone® RL, suitable for microbiology