MilliporeSigma
  • Comparative efficacy of several disinfectants in suspension and carrier tests against Haemophilus parasuis serovars 1 and 5.
Comparative efficacy of several disinfectants in suspension and carrier tests against Haemophilus parasuis serovars 1 and 5.
Research in veterinary science (2009-12-29)
E F Rodríguez Ferri, S Martínez, R Frandoloso, S Yubero, C B Gutiérrez Martín
ABSTRACT
The comparative efficacy of 16 active compounds (including the most commonly used chemical groups) and 10 commercial formulations against Haemophilus parasuis serovars 1 and 5 was studied. These organisms were tested in suspension and carrier tests in the presence and absence of serum as representative of organic matter. Chloramine-T and half of the formulations from commercial sources (most of them including quaternary ammonium compounds) were effective in both in vitro tests, regardless of the presence or absence of organic load. All 26 disinfectants except for an iodophor (0.1% available iodine) resulted in at least 3-log(10) reduction in colony-forming units in suspension test, and most of them resulted in the maximal level of detection (>6-log(10) reduction). On the other hand, disinfectants were not as effective in carrier test as in suspension test, and the presence of serum considerably reduced the activities of most of the compounds tested, especially in carrier test. These results suggest the importance of selecting suitable disinfection for routine use on surfaces contaminated with H. parasuis, particularly when organic matter is present. Chloramine-T and formulations 2 and 7-10 are recommended for a complete inactivation of H. parasuis in swine herds.
Sigma-Aldrich
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis