MilliporeSigma
  • One-year treatment with 0.1% tacrolimus ointment versus a corticosteroid regimen in adults with moderate to severe atopic dermatitis: A randomized, double-blind, comparative trial.
One-year treatment with 0.1% tacrolimus ointment versus a corticosteroid regimen in adults with moderate to severe atopic dermatitis: A randomized, double-blind, comparative trial.
Acta dermato-venereologica (2010-02-20)
Johanna Mandelin, Anita Remitz, Hannele Virtanen, Sakari Reitamo
ABSTRACT
A one-year, randomized, double-blind study was conducted in 80 patients with atopic dermatitis treated with tacrolimus ointment or a corticosteroid regimen (hydrocortisone acetate 1% ointment for head and neck, hydrocortisone butyrate 0.1% ointment for trunk and limbs) to compare efficacy and safety, and effects on Th2-reactivity. The study was completed by 36/40 patients in the tacrolimus group, and 31/40 patients in the corticosteroid group. In both groups affected body surface area, eczema area and severity index, and transepidermal water loss decreased at months 6 and 12. Tacrolimus was superior for all efficacy scores at month 6, and in the head and neck area at month 12. Recall antigen reactivity increased at month 12 in both groups. Adverse events were reported by 40/40 patients in the tacrolimus, and by 34/40 patients in the corticosteroid group. Long-term treatment with topical tacrolimus or a corticosteroid regimen improves atopic dermatitis and recall antigen reactivity, suggesting an improvement in the Th1/Th2-balance.
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications
Hydrocortisone acetate, European Pharmacopoeia (EP) Reference Standard