MilliporeSigma
  • Transient state monitoring by total internal reflection fluorescence microscopy.
Transient state monitoring by total internal reflection fluorescence microscopy.
The journal of physical chemistry. B (2010-03-04)
Thiemo Spielmann, Hans Blom, Matthias Geissbuehler, Theo Lasser, Jerker Widengren
ABSTRACT
Triplet, photo-oxidized and other photoinduced, long-lived states of fluorophores are sensitive to the local environment and thus attractive for microenvironmental imaging purposes. In this work, we introduce an approach where these states are monitored in a total internal reflection (TIR) fluorescence microscope, via the characteristic variations of the time-averaged fluorescence occurring in response to different excitation modulation schemes. The surface-confined TIR excitation field generates a signal from the fluorescent molecules close to the glass surface. Thereby, a high selectivity and low background noise is obtained, and in combination with low duty cycles of excitation, the overall photodegradation of the fluorescent molecules of the sample can be kept low. To verify the approach, the kinetics of the triplet and radical states of the dye Rhodamine 110 were imaged and analyzed in aqueous solutions at different concentrations of dissolved oxygen and of the reducing agent ascorbic acid. The experimental results were compared to data from corresponding fluorescence correlation spectroscopy (FCS) measurements and simulations based on finite element analysis. The approach was found to accurately determine relative populations and dynamics of triplet and photo-oxidized states, overcoming passage time limitations seen in FCS measurements. The method circumvents the need for time resolution in the fluorescence detection, allowing simultaneous readout over the whole surface area subject to excitation. It can be applied over a broad range of concentrations and does not require a strong fluorescence brightness of the sample molecules. Given the sensitivity of the triplet and photo-oxidized states to oxygen concentrations and not the least to local redox environments, we expect the approach to become an attractive tool for imaging cell metabolism.
Sigma-Aldrich
Rhodamine 110 chloride, suitable for fluorescence, BioReagent, ≥99.0% (UV)
Sigma-Aldrich
Rhodamine 110 chloride, Dye content ≥75 %