MilliporeSigma
  • Direct evidence of chloride ion efflux in ischaemic and pharmacological preconditioning of cultured cardiomyocytes.
Direct evidence of chloride ion efflux in ischaemic and pharmacological preconditioning of cultured cardiomyocytes.
Cardiovascular research (2010-03-17)
Roberto J Diaz, Alina Hinek, Gregory J Wilson
ABSTRACT
We have previously shown that reduction of ischaemic cell swelling via enhanced cell volume regulation is a key mechanism of ischaemic preconditioning (IPC) in cardiomyocytes. We have also shown that pharmacological blockade of Cl(-) channels abolishes cardioprotection achieved by IPC in Langendorff-perfused hearts and freshly isolated cardiomyocytes, thus suggesting that Cl(-) plays a key role in IPC cardioprotection. However, direct evidence of Cl(-) channel activation resulting in transsarcolemmal Cl(-) efflux by IPC had been lacking. To address this issue, 24 h cultured rabbit cardiomyocytes were loaded with 5 mM 6-methoxy-N-(3-sulfopropyl)quinolinium (SPQ), a specific fluorescence indicator that is quenched by Cl(-) so that cellular efflux of Cl(-) results in an increase in SPQ fluorescence. After stabilization for 10 min, cardiomyocytes were preconditioned either with 10 min simulated ischaemia/10 min simulated reperfusion or with 10 min treatment with 1 microM N(6)-2-(4-aminophenyl)ethyladenosine (APNEA). IPC and APNEA significantly (P < 0.001) reduced the intracellular Cl(-) concentration ([Cl(-)](i)) to 31.9 +/- 3.2 mM (mean +/- SEM) and 32.5 +/- 2.8 mM, respectively, from an initial [Cl(-)](i) (pooled stabilization 61.5 +/- 7.1 mM). [Cl(-)](i) did not change in control (non-preconditioned) cardiomyocytes (control 58.1 +/- 1.9 mM and control + vehicle 62.6 +/- 4.9 mM, P = 0.98 and 0.99 vs. pooled pre-treatment baseline, respectively). Inhibition of Cl(-) channels with 50 microM indanyloxyacetic acid 94 completely blocked preconditioning-induced Cl(-) efflux. Thus, a net Cl(-) efflux of 29.6 and 29.0 mM was triggered by IPC and APNEA. These findings provide the first direct evidence of activation of sarcolemmal Cl(-) channels by ischaemic and pharmacological preconditioning in cardiomyocytes.
Sigma-Aldrich
R(+)-IAA-94, ≥98% (HPLC), solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685