MilliporeSigma
  • Validation of an isocratic HPLC method to detect 2-fluoro-beta-alanine for the analysis of dihydropyrimidine dehydrogenase activity.
Validation of an isocratic HPLC method to detect 2-fluoro-beta-alanine for the analysis of dihydropyrimidine dehydrogenase activity.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-06-24)
Kinta M Serve, Jennifer L Darnell, Jody K Takemoto, Neal M Davies, Margaret E Black
ABSTRACT
The efficacy of the chemotherapeutic drug 5'-fluorouracil is reduced by catabolism to 2'-fluoro-beta-alanine (FBAL), a three-step reaction in which dihydropyrimidine dehydrogenase (DPD) catalyzes the rate-limiting step. To study in vitro DPD activity, we developed and validated an isocratic, reverse-phase HPLC method to detect and quantify FBAL without using multiple columns or radiolabeled substrates. Pre-column derivatization of FBAL was performed using o-phthalaldehyde in the presence of two sulfur donors, ethanthiol or beta-mercaptoethanol, and the resulting products assayed. Calibration curves were linear over a range of 10-200 microg/ml and the method was successfully applied to the examination of DPD activity in cultured cells.
Sigma-Aldrich
Ethanethiol, 97%
Sigma-Aldrich
Ethanethiol, ≥97%
Sigma-Aldrich
Sodium ethanethiolate, technical grade, 80%
Sigma-Aldrich
Sodium ethanethiolate, technical, ~90% (RT)
Sigma-Aldrich
3-Amino-2-fluoropropionic acid, 95%